Assay Details
| Gene Symbol: Gm31135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK082314.1 | - | 357 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 10, member 2 |
Slc10a2 |
ASBT, ISBT |
-3.0mb | Sense | Chr. 8 - 5085623 - 5105232 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 31343 |
Gm31343 | 236.9kb | Anti-sense | Chr. 8 - 8377090 - 8391702 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm31135
Genomic Map