Assay Details
Gene Symbol: Nap1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138742.1 | NP_620081.1 | 1 | 2237 | 100 | |
GenBank mRNA | AB010711.1 | - | 1 | 2253 | 100 | |
AK078521.1 | - | 1 | 2229 | 100 | ||
BC020176.1 | - | 1 | 2237 | 4505496 | 100 | |
BC027045.1 | - | 1 | 2134 | 5363560 | 100 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213928 | BC027045.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213928 | AK078521.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213928 | BC020176.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213928 | NM_138742.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213928 | AB010711.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213928 | BC027045.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213928 | AK078521.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213928 | BC020176.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213928 | NM_138742.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213928 | AB010711.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367161 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 645268 | AK053018.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 645268 | NM_183185.3 | refseq_mrna_mouse_NM |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108600 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213928 | BC027045.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213928 | AK078521.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213928 | BC020176.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213928 | NM_138742.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213928 | AB010711.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213928 | BC027045.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213928 | AK078521.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213928 | BC020176.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213928 | NM_138742.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213928 | AB010711.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P213928 | BC027045.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P213928 | AK078521.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P213928 | BC020176.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P213928 | NM_138742.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P213928 | AB010711.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P213928 | BC027045.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P213928 | AK078521.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P213928 | BC020176.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P213928 | NM_138742.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P213928 | AB010711.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632418 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632419 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632427 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632431 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632440 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632450 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632416 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632468 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632460 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632442 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632433 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632452 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632459 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632455 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632453 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632439 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632446 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632415 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632414 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632430 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632458 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632424 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632423 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632456 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632435 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632462 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632461 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632444 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632421 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632436 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632464 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632443 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632448 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632441 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632428 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32632465 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108600 | - | - |
Whole Mouse Genome G4122A | A_52_P500337 | BC027045.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P500337 | AK078521.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P500337 | BC020176.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P500337 | NM_138742.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P500337 | AB010711.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P213928 | BC027045.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P213928 | AK078521.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P213928 | BC020176.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P213928 | NM_138742.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P213928 | AB010711.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P213928 | BC027045.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P213928 | AK078521.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P213928 | BC020176.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P213928 | NM_138742.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P213928 | AB010711.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112746 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128142 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108600 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15128729 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15128730 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15128728 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15128732 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15128723 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15128710 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14542783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632433 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632402 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632444 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632456 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632415 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32632373 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632470 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632419 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632471 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632452 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14542783 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632462 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632413 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14610641 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632441 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632424 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632396 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632455 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632406 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632448 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632435 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632456 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632465 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632458 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632450 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632461 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632418 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15001427 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632407 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632402 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632442 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632460 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632427 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632412 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632415 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632430 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632443 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632398 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632436 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632446 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632453 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632423 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632459 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632431 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632439 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632421 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632468 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632408 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632466 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14597348 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32632444 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14976657 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10606558 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10606555 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10606556 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10606557 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418500_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10606554 | - | - |
Mouse Genome 430 2.0 Array | 1418500_at | - | - |
Mouse Genome 430 2.0 Array | 1418500_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54561_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54561_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0X00018396.mm.1 | NM_138742.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00018396.mm.1 | NM_138742.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00018396.mm.1 | TC0X00002919.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00018395.mm.1 | NM_138742.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00018395.mm.1 | NM_138742.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00018395.mm.1 | TC0X00002919.mm.1 | MTA TC |
Mouse Expression Set 430 | 1418500_PM_at | - | - |
Mouse Expression Set 430 | 1418500_at | - | - |
Murine Genome U74v2 Set | 166171_at | - | - |
Murine Genome U74v2 Set | 166171_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418500_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418500_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418500_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418500_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 642710 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6020079 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6020079 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006020079 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1217839 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1217839 | - | - |
mouse-6 v1.1 (Illumina) | scl54673.1_134-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6020079 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl54673.1_134-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6020079 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1217839 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1217839 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl54673.1_134-S | - | - |
