Assay Details
| Gene Symbol: Rab11fip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029423.2 | NP_083699.2 | 3 - 4 | 1671 | 93 | |
| GenBank mRNA | AK014696.1 | - | 3 - 4 | 1653 | 93 | |
| AK030769.1 | - | 3 - 4 | 1674 | 93 | ||
| AK151024.1 | - | 3 - 4 | 1672 | 93 | ||
| BC125400.1 | - | 3 - 4 | 1656 | 40129915 | 93 | |
| BC132094.1 | - | 3 - 4 | 1656 | 40130371 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab11fip1
Genomic Map