Assay Details
Gene Symbol: Prosc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039077.2 | NP_001034166.1 | 1 - 2 | 159 | 93 | |
NM_001039078.2 | NP_001034167.1 | 1 - 2 | 159 | 93 | ||
NM_054057.4 | NP_473398.1 | 1 - 2 | 159 | 93 | ||
GenBank mRNA | AK032308.1 | - | 1 - 2 | 109 | 93 | |
AK049905.1 | - | 1 - 2 | 118 | 93 | ||
AK134549.1 | - | 1 - 2 | 92 | 93 | ||
AK147670.1 | - | 1 - 2 | 141 | 93 | ||
BC047992.1 | - | 1 - 2 | 116 | 6504579 | 93 | |
BC061045.1 | - | 1 - 2 | 153 | 6534037 | 93 | |
BY065041.1 | - | 1 - 2 | 159 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
