Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800002642.mm.1 |
NM_001039077.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002642.mm.1 |
NM_001039077.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002642.mm.1 |
NM_001039078.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002642.mm.1 |
NM_001039078.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002642.mm.1 |
NM_054057.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002642.mm.1 |
NM_054057.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002642.mm.1 |
TC0800000346.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800002645.mm.1 |
NM_001039077.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002645.mm.1 |
NM_001039077.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002645.mm.1 |
NM_054057.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002645.mm.1 |
NM_054057.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002645.mm.1 |
TC0800000346.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800002644.mm.1 |
NM_001039077.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002644.mm.1 |
NM_001039077.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002644.mm.1 |
NM_001039078.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002644.mm.1 |
NM_001039078.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002644.mm.1 |
NM_054057.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002644.mm.1 |
NM_054057.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002644.mm.1 |
TC0800000346.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800002643.mm.1 |
NM_001039077.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002643.mm.1 |
NM_001039077.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002643.mm.1 |
NM_001039078.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002643.mm.1 |
NM_001039078.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002643.mm.1 |
NM_054057.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002643.mm.1 |
NM_054057.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002643.mm.1 |
TC0800000346.mm.1 |
MTA TC |