Assay Details
Gene Symbol: Prosc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054057.4 | NP_473398.1 | 5 - 6 | 517 | 80 | |
GenBank mRNA | AK032308.1 | - | 5 - 6 | 467 | 80 | |
AK160172.1 | - | 5 - 6 | 471 | 80 | ||
AK213501.1 | - | 2 - 3 | 155 | 80 | ||
AK218333.1 | - | 2 - 3 | 156 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
