Assay Details
Gene Symbol: Prosc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054057.4 | NP_473398.1 | 7 - 8 | 751 | 74 | |
GenBank mRNA | AK032308.1 | - | 7 - 8 | 701 | 74 | |
AK160172.1 | - | 7 - 8 | 705 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
