Assay Details
| Gene Symbol: Rbbp8nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173031.3 | NP_766619.2 | 4 - 5 | 377 | 71 | |
| GenBank mRNA | BC066135.1 | - | 4 - 5 | 333 | 71 | |
| BC150940.1 | - | 4 - 5 | 332 | 71 | ||
| BC172093.1 | - | 4 - 5 | 332 | 71 | ||
| BY714078.1 | - | 4 - 5 | 379 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbbp8nl
Genomic Map