Assay Details
| Gene Symbol: Helz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183162.2 | NP_898985.2 | 6 - 7 | 2600 | 72 | |
| GenBank mRNA | AB201714.1 | - | 5 - 6 | 2504 | 72 | |
| AK157942.1 | - | 6 - 7 | 2599 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Helz2
Genomic Map