Assay Details
| Gene Symbol: Dido1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291433.1 | NP_001278362.1 | 9 - 10 | 2654 | 153 | |
| NM_175551.4 | NP_780760.2 | 9 - 10 | 2654 | 153 | ||
| NM_177852.4 | NP_808520.2 | 9 - 10 | 2602 | 153 | ||
| NR_111965.1 | 7 - 8 | 2352 | 153 | |||
| GenBank mRNA | AK049707.1 | - | 3 - 4 | 1269 | 153 | |
| AK129117.1 | - | 9 - 10 | 2602 | 153 | ||
| AY425951.1 | - | 9 - 10 | 2556 | 153 | ||
| AY425952.1 | - | 9 - 10 | 2556 | 153 | ||
| BC044755.1 | - | 3 - 4 | 753 | 153 | ||
| BC138712.1 | - | 8 - 9 | 2431 | 8861734 | 153 | |
| BC138713.1 | - | 8 - 9 | 2431 | 8861735 | 153 | |
| BC145401.1 | - | 7 - 8 | 2272 | 153 | ||
More Information
Set Membership: |
|

Dido1
Genomic Map