Assay Details
| Gene Symbol: Mrgbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028479.1 | NP_082755.1 | 2 - 3 | 575 | 83 | |
| GenBank mRNA | AK005551.1 | - | 1 - 2 | 215 | 83 | |
| AK052575.1 | - | 1 - 2 | 230 | 83 | ||
| BC042508.1 | - | 1 - 2 | 189 | 83 | ||
| BC054431.1 | - | 1 - 2 | 162 | 83 | ||
| BC110993.1 | - | 1 - 2 | 184 | 5363496 | 83 | |
| BC116732.1 | - | 2 - 3 | 575 | 40126051 | 83 | |
| BC116734.1 | - | 2 - 3 | 575 | 40126053 | 83 | |
More Information
Set Membership: |
|

Mrgbp
Genomic Map