Assay Details
| Gene Symbol: Mrgbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028479.1 | NP_082755.1 | 4 - 5 | 787 | 127 | |
| GenBank mRNA | AK005551.1 | - | 3 - 4 | 427 | 127 | |
| AK217601.1 | - | 2 - 3 | 204 | 127 | ||
| BC110993.1 | - | 3 - 4 | 393 | 5363496 | 127 | |
| BC116732.1 | - | 4 - 5 | 787 | 40126051 | 127 | |
| BC116734.1 | - | 4 - 5 | 787 | 40126053 | 127 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mrgbp
Genomic Map