Assay Details
| Gene Symbol: Mrgbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028479.1 | NP_082755.1 | 5 - 6 | 858 | 126 | |
| GenBank mRNA | AK005551.1 | - | 4 - 5 | 498 | 126 | |
| AK038431.1 | - | 3 - 4 | 485 | 126 | ||
| AK217601.1 | - | 3 - 4 | 275 | 126 | ||
| BC042508.1 | - | 2 - 3 | 1664 | 126 | ||
| BC054431.1 | - | 3 - 4 | 754 | 126 | ||
| BC085320.1 | - | 1 - 2 | 2157 | 126 | ||
| BC110993.1 | - | 4 - 5 | 464 | 5363496 | 126 | |
| BC116732.1 | - | 5 - 6 | 858 | 40126051 | 126 | |
| BC116734.1 | - | 5 - 6 | 858 | 40126053 | 126 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21904831 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21904830 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21904833 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171993 | BC116734.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171993 | BC085320.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171993 | BC110993.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171993 | BC042508.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171993 | BC054431.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 420010 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 255902 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 796890 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133078 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171993 | BC116734.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171993 | BC085320.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171993 | BC110993.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171993 | BC042508.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171993 | BC054431.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133078 | - | - |
| Whole Mouse Genome G4122A | A_51_P422691 | BC085320.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422691 | BC110993.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422691 | BC042508.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422691 | BC054431.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422691 | AK005551.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133078 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21904831 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21904830 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21904833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21904899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01551991 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01601142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01552003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904806 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01374556 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904847 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02046567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01601149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904816 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02046563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21904792 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10479405 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10479404 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10479407 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10479409 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10479408 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10479403 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10479406 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436418_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428366_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10479402 | - | - |
| Mouse Genome 430 2.0 Array | 1428366_at | - | - |
| Mouse Genome 430 2.0 Array | 1436418_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436418_at | - | - |
| Mouse Genome 430 2.0 Array | 1428366_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200022868.mm.1 | TC0200002794.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200022866.mm.1 | NM_028479.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200022866.mm.1 | NM_028479.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200022866.mm.1 | TC0200002794.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436418_PM_at | - | - |
| Mouse Expression Set 430 | 1428366_at | - | - |
| Mouse Expression Set 430 | 1436418_at | - | - |
| Mouse Expression Set 430 | 1428366_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436418_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428366_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436418_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428366_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 103060019 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257064 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103060019 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103060019 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2805631 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257064 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2805631 | - | - |

Mrgbp
Genomic Map