| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200022851.mm.1 |
NM_028479.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022851.mm.1 |
NM_028479.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022851.mm.1 |
TC0200002794.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200022856.mm.1 |
NM_028479.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022856.mm.1 |
NM_028479.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022856.mm.1 |
TC0200002794.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200022852.mm.1 |
NM_028479.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022852.mm.1 |
NM_028479.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022852.mm.1 |
TC0200002794.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200022858.mm.1 |
NM_028479.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022858.mm.1 |
NM_028479.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022858.mm.1 |
TC0200002794.mm.1 |
MTA TC |