Assay Details
| Gene Symbol: Lsm14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177727.4 | NP_808395.2 | 5 - 6 | 870 | 79 | |
| GenBank mRNA | AK035040.1 | - | 5 - 6 | 899 | 79 | |
| BC040823.1 | - | 5 - 6 | 870 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lsm14b
Genomic Map