Assay Details
| Gene Symbol: Tpd52l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291201.1 | NP_001278130.1 | 3 - 4 | 489 | 64 | |
| NM_001291202.1 | NP_001278131.1 | 3 - 4 | 489 | 64 | ||
| NM_001291203.1 | NP_001278132.1 | 3 - 4 | 489 | 64 | ||
| GenBank mRNA | AK051268.1 | - | 3 - 4 | 433 | 64 | |
| AK155987.1 | - | 3 - 4 | 426 | 64 | ||
| AK160727.1 | - | 3 - 4 | 433 | 64 | ||
| AK161092.1 | - | 3 - 4 | 445 | 64 | ||
More Information
Set Membership: |
|

Tpd52l2
Genomic Map