Assay Details
| Gene Symbol: Tpd52l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291197.1 | NP_001278126.1 | 4 - 5 | 543 | 83 | |
| NM_001291200.1 | NP_001278129.1 | 4 - 5 | 543 | 83 | ||
| NM_025482.3 | NP_079758.2 | 4 - 5 | 543 | 83 | ||
| NR_111899.1 | 3 - 4 | 397 | 83 | |||
| GenBank mRNA | AK013085.1 | - | 4 - 5 | 485 | 83 | |
| AK013188.1 | - | 4 - 5 | 496 | 83 | ||
| AK147885.1 | - | 4 - 5 | 503 | 83 | ||
| AK159925.1 | - | 4 - 5 | 471 | 83 | ||
| AK171825.1 | - | 4 - 5 | 505 | 83 | ||
| BC006886.1 | - | 4 - 5 | 432 | 2647702 | 83 | |
| CN721218.1 | - | 3 - 4 | 397 | 83 | ||
| DV047279.1 | - | 4 - 5 | 529 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tpd52l2
Genomic Map