Assay Details
| Gene Symbol: Dnajc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271584.1 | NP_001258513.1 | 2 - 3 | 271 | 117 | |
| GenBank mRNA | BC012268.1 | - | 2 - 3 | 188 | 3500124 | 119 |
| BY101584.1 | - | 2 - 3 | 206 | 117 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC012268.1, NM_001271584.1 transcripts.
Set Membership: |
|

Dnajc5
Genomic Map