Assay Details
| Gene Symbol: Dnajc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073369.1 | 5 - 6 | 792 | 62 | ||
| GenBank mRNA | AK007879.1 | - | 5 - 6 | 734 | 62 | |
| AK083584.1 | - | 5 - 6 | 754 | 62 | ||
| CN459638.1 | - | 5 - 6 | 608 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnajc5
Genomic Map