Assay Details
Gene Symbol: Itgb2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008405.3 | NP_032431.2 | 9 - 10 | 1078 | 73 | |
GenBank mRNA | AF051367.1 | - | 9 - 10 | 1063 | 73 | |
AK036640.1 | - | 9 - 10 | 1078 | 73 | ||
AK137534.1 | - | 9 - 10 | 1079 | 73 | ||
AK158108.1 | - | 9 - 10 | 1079 | 73 | ||
BC113142.1 | - | 9 - 10 | 1047 | 40047633 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
