Assay Details
| Gene Symbol: Tmprss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015775.2 | NP_056590.2 | 12 - 13 | 1533 | 91 | |
| GenBank mRNA | AF113596.1 | - | 12 - 13 | 1403 | 91 | |
| AF199362.1 | - | 12 - 13 | 1414 | 91 | ||
| AF243500.1 | - | 12 - 13 | 1403 | 91 | ||
| AK141350.1 | - | 12 - 13 | 1508 | 91 | ||
| AK146047.1 | - | 12 - 13 | 1417 | 91 | ||
| BC038393.1 | - | 12 - 13 | 1372 | 2648878 | 91 | |
| BC054348.1 | - | 12 - 13 | 1533 | 2648889 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss2
Genomic Map