Assay Details
| Gene Symbol: Tmprss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015775.2 | NP_056590.2 | 1 - 2 | 176 | 73 | |
| GenBank mRNA | AF199362.1 | - | 1 - 2 | 57 | 73 | |
| AK141350.1 | - | 1 - 2 | 150 | 73 | ||
| AK146047.1 | - | 1 - 2 | 60 | 73 | ||
| AK161173.1 | - | 1 - 2 | 170 | 73 | ||
| BC054348.1 | - | 1 - 2 | 176 | 2648889 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF199362.1, AK146047.1, AK161173.1, BC054348.1, NM_015775.2 transcripts.
Set Membership: |
|

Tmprss2
Genomic Map