Assay Details
| Gene Symbol: Kdm5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011419.3 | NP_035549.1 | 13 - 14 | 2040 | 61 | |
| GenBank mRNA | AB221607.1 | - | 13 - 14 | 1866 | 61 | |
| AF127244.1 | - | 13 - 14 | 1878 | 61 | ||
| AK140971.1 | - | 13 - 14 | 2035 | 61 | ||
| BC059077.1 | - | 5 - 6 | 855 | 6826863 | 61 | |
| BC060077.1 | - | 2019 | 61 | |||
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0Y00000005.rn
Set Membership: |
|

Kdm5d
Genomic Map