Assay Details
| Gene Symbol: Kdm5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011419.3 | NP_035549.1 | 2 - 3 | 406 | 133 | |
| GenBank mRNA | AB221607.1 | - | 2 - 3 | 232 | 133 | |
| AF127244.1 | - | 2 - 3 | 244 | 133 | ||
| AK047790.1 | - | 2 - 3 | 407 | 133 | ||
| AK140971.1 | - | 2 - 3 | 401 | 133 | ||
| AK141096.1 | - | 2 - 3 | 402 | 133 | ||
| BB647801.1 | - | 2 - 3 | 404 | 133 | ||
| BC060077.1 | - | 356 | 133 | |||
| Z29652.1 | - | 2 - 3 | 162 | 133 | ||
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0Y00000005.rn
Set Membership: |
|

Kdm5d
Genomic Map