Assay Details
| Gene Symbol: Kdm5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011419.3 | NP_035549.1 | 8 - 9 | 1298 | 70 | |
| GenBank mRNA | AB221607.1 | - | 8 - 9 | 1124 | 70 | |
| AF127244.1 | - | 8 - 9 | 1136 | 70 | ||
| AK047790.1 | - | 8 - 9 | 1299 | 70 | ||
| AK140971.1 | - | 8 - 9 | 1293 | 70 | ||
| BC060077.1 | - | 1277 | 70 | |||
| Z29652.1 | - | 8 - 9 | 1054 | 70 | ||
More Information
Set Membership: |
|

Kdm5d
Genomic Map