Assay Details
| Gene Symbol: Il17rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019583.3 | NP_062529.2 | 2 - 3 | 189 | 74 | |
| GenBank mRNA | AF208108.1 | - | 2 - 3 | 102 | 74 | |
| AF208109.1 | - | 2 - 3 | 102 | 74 | ||
| AK045627.1 | - | 2 - 3 | 190 | 74 | ||
| AK050139.1 | - | 2 - 3 | 106 | 74 | ||
| BC026546.1 | - | 2 - 3 | 89 | 5042466 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il17rb
Genomic Map