Assay Details
| Gene Symbol: Il17rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019583.3 | NP_062529.2 | 5 - 6 | 586 | 79 | |
| GenBank mRNA | AF208108.1 | - | 5 - 6 | 499 | 79 | |
| AK045627.1 | - | 5 - 6 | 587 | 79 | ||
| AK050139.1 | - | 5 - 6 | 507 | 79 | ||
| BC026546.1 | - | 5 - 6 | 486 | 5042466 | 79 | |
More Information
Set Membership: |
|

Il17rb
Genomic Map