Assay Details
| Gene Symbol: Nek5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177898.4 | NP_808566.2 | 12 - 13 | 1156 | 82 | |
| GenBank mRNA | AK032672.1 | - | 12 - 13 | 1158 | 82 | |
| AK165108.1 | - | 10 - 11 | 905 | 82 | ||
| BC053516.1 | - | 12 - 13 | 1144 | 6311513 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nek5
Genomic Map