Assay Details
| Gene Symbol: Nek5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177898.4 | NP_808566.2 | 2 - 3 | 207 | 94 | |
| GenBank mRNA | AK032672.1 | - | 2 - 3 | 209 | 94 | |
| AK054168.1 | - | 2 - 3 | 221 | 94 | ||
| BC053516.1 | - | 2 - 3 | 195 | 6311513 | 94 | |
More Information
Set Membership: |
|

Nek5
Genomic Map