Assay Details
| Gene Symbol: Nek5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177898.4 | NP_808566.2 | 5 - 6 | 466 | 125 | |
| GenBank mRNA | AK032672.1 | - | 5 - 6 | 468 | 125 | |
| AK054168.1 | - | 5 - 6 | 480 | 125 | ||
| AK165108.1 | - | 3 - 4 | 229 | 125 | ||
| BC053516.1 | - | 5 - 6 | 454 | 6311513 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nek5
Genomic Map