Assay Details
| Gene Symbol: Gart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010256.2 | NP_034386.2 | 1 - 2 | 90 | 56 | |
| GenBank mRNA | AK146355.1 | - | 1 - 2 | 75 | 56 | |
| AK168501.1 | - | 1 - 2 | 53 | 56 | ||
| AK168724.1 | - | 1 - 2 | 65 | 56 | ||
| AK168796.1 | - | 1 - 2 | 65 | 56 | ||
| AK168876.1 | - | 1 - 2 | 42 | 56 | ||
| BB843046.1 | - | 1 - 2 | 91 | 56 | ||
| BC070465.1 | - | 1 - 2 | 37 | 30611835 | 56 | |
| U01023.1 | - | 1 - 2 | 61 | 56 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK146355.1, AK168501.1, AK168724.1, AK168796.1, AK168876.1, BC070465.1, NM_010256.2, U01023.1 transcripts.
Set Membership: |
|

Gart
Genomic Map