Assay Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009441.2 | NP_033467.2 | 7 - 8 | 726 | 88 | |
| GenBank mRNA | AB008516.1 | - | 6 - 7 | 603 | 88 | |
| AK053765.1 | - | 7 - 8 | 830 | 88 | ||
| AK088273.1 | - | 6 - 7 | 673 | 88 | ||
| AK166298.1 | - | 7 - 8 | 668 | 88 | ||
| AK170087.1 | - | 6 - 7 | 674 | 88 | ||
| AK220495.1 | - | 7 - 8 | 768 | 88 | ||
| BC019173.1 | - | 7 - 8 | 695 | 88 | ||
| BC037654.1 | - | 6 - 7 | 628 | 88 | ||
| BC049277.1 | - | 6 - 7 | 665 | 88 | ||
| BC057207.1 | - | 6 - 7 | 630 | 88 | ||
| BC145489.1 | - | 6 - 7 | 643 | 88 | ||
More Information
Set Membership: |
|

Ttc3
Genomic Map