Assay Details
| Gene Symbol: Dscr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007834.3 | NP_031860.1 | 6 - 7 | 890 | 67 | |
| GenBank mRNA | AB001990.1 | - | 6 - 7 | 705 | 67 | |
| AK077485.1 | - | 6 - 7 | 704 | 67 | ||
| AK077628.1 | - | 6 - 7 | 696 | 67 | ||
| BC003740.1 | - | 6 - 7 | 678 | 3592608 | 67 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dscr3
Genomic Map