Assay Details
| Gene Symbol: Pigp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159616.1 | NP_001153088.1 | 1 - 2 | 104 | 117 | |
| GenBank mRNA | AF216306.1 | - | 1 - 2 | 106 | 117 | |
| BC096569.1 | - | 1 - 2 | 111 | 1429742 | 117 | |
| BY041400.1 | - | 1 - 2 | 104 | 117 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF216306.1,BC096569.1,NM_001159616.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pigp
Genomic Map