Assay Details
| Gene Symbol: Kcnj6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025590.1 | NP_001020761.1 | 2 - 3 | 1153 | 64 | |
| GenBank mRNA | U51126.1 | - | 2 - 3 | 1141 | 64 | |
More Information
Set Membership: |
|

Kcnj6
Genomic Map