Assay Details
| Gene Symbol: F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161373.1 | NP_001154845.1 | 11 - 12 | 2156 | 124 | |
| NM_001161374.1 | NP_001154846.1 | 11 - 12 | 2156 | 124 | ||
| NM_007977.2 | NP_032003.2 | 11 - 12 | 2156 | 124 | ||
| GenBank mRNA | BC138347.1 | - | 11 - 12 | 2037 | 8861367 | 124 |
| BC145245.1 | - | 11 - 12 | 2037 | 9053797 | 124 | |
| L05573.1 | - | 11 - 12 | 2154 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

F8
Genomic Map