Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00015796.mm.1 |
NM_001161373.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015796.mm.1 |
NM_001161373.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015796.mm.1 |
NM_001161374.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015796.mm.1 |
NM_001161374.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015796.mm.1 |
NM_007977.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015796.mm.1 |
NM_007977.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015796.mm.1 |
TC0X00002472.mm.1 |
MTA TC |