Assay Details
| Gene Symbol: Pitpnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008851.4 | NP_032877.1 | 19 - 20 | 3561 | 70 | |
| NR_075078.1 | 20 - 21 | 3255 | 70 | |||
| GenBank mRNA | AF006467.1 | - | 20 - 21 | 3136 | 70 | |
| BC044893.1 | - | 19 - 20 | 3066 | 4481550 | 70 | |
| BC048150.1 | - | 19 - 20 | 3215 | 5706382 | 70 | |
| Y08922.1 | - | 19 - 20 | 3562 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pitpnm1
Genomic Map