Assay Details
| Gene Symbol: Pitpnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008851.4 | NP_032877.1 | 21 - 22 | 3909 | 85 | |
| NR_075078.1 | 22 - 23 | 3603 | 85 | |||
| GenBank mRNA | AF006467.1 | - | 22 - 23 | 3484 | 85 | |
| AK155377.1 | - | 21 - 22 | 4382 | 85 | ||
| AK207185.1 | - | 1 - 2 | 51 | 85 | ||
| BC044893.1 | - | 21 - 22 | 3414 | 4481550 | 85 | |
| BC048150.1 | - | 21 - 22 | 3563 | 5706382 | 85 | |
| Y08922.1 | - | 21 - 22 | 3910 | 85 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pitpnm1
Genomic Map