| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900000343.mm.1 |
NM_008851.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000343.mm.1 |
NR_075078.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000343.mm.1 |
TC1900000027.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900000340.mm.1 |
NM_008851.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000340.mm.1 |
TC1900000027.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900000339.mm.1 |
NM_008851.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000339.mm.1 |
TC1900000027.mm.1 |
MTA TC |