Assay Details
| Gene Symbol: Slc34a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011402.3 | NP_035532.2 | 5 - 6 | 574 | 84 | |
| GenBank mRNA | AF081499.1 | - | 5 - 6 | 571 | 84 | |
| AK004832.1 | - | 5 - 6 | 573 | 84 | ||
| AK145599.1 | - | 5 - 6 | 573 | 84 | ||
| AK164786.1 | - | 5 - 6 | 575 | 84 | ||
| BC096369.1 | - | 4 - 5 | 545 | 3962784 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc34a2
Genomic Map