Assay Details
| Gene Symbol: Sel1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172710.3 | NP_766298.2 | 8 - 9 | 1587 | 76 | |
| GenBank mRNA | AK030487.1 | - | 8 - 9 | 1452 | 76 | |
| AK037220.1 | - | 8 - 9 | 1451 | 76 | ||
| AK122362.1 | - | 8 - 9 | 1587 | 76 | ||
| BC094607.1 | - | 7 - 8 | 1132 | 6837048 | 76 | |
| BC117915.1 | - | 7 - 8 | 1088 | 40097358 | 76 | |
| BC117916.1 | - | 7 - 8 | 1090 | 40097359 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sel1l3
Genomic Map