Assay Details
| Gene Symbol: Dlgap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145965.1 | NP_001139437.1 | 4 - 5 | 420 | 56 | |
| NM_172910.3 | NP_766498.2 | 5 - 6 | 519 | 56 | ||
| GenBank mRNA | AK032564.1 | - | 5 - 6 | 520 | 56 | |
| AK082945.1 | - | 3 - 4 | 292 | 56 | ||
| BB622956.1 | - | 520 | 56 | |||
| BC118969.2 | - | 4 - 5 | 206 | 40097764 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dlgap2
Genomic Map