| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500021156.mm.1 |
TC0500002290.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021156.mm.1 |
NM_013748.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021156.mm.1 |
NM_013748.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021160.mm.1 |
TC0500002290.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021160.mm.1 |
NM_013748.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021160.mm.1 |
NM_013748.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021155.mm.1 |
TC0500002290.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021155.mm.1 |
NM_013748.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021155.mm.1 |
NM_013748.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021157.mm.1 |
TC0500002290.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021157.mm.1 |
NM_013748.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021157.mm.1 |
NM_013748.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021158.mm.1 |
TC0500002290.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021158.mm.1 |
NM_013748.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021158.mm.1 |
NM_013748.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021159.mm.1 |
TC0500002290.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021159.mm.1 |
NM_013748.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021159.mm.1 |
NM_013748.3 |
Refseq |