Assay Details
| Gene Symbol: Preb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294302.1 | NP_001281231.1 | 5 - 6 | 973 | 69 | |
| NM_016703.3 | NP_057912.2 | 5 - 6 | 973 | 69 | ||
| GenBank mRNA | AF150808.1 | - | 5 - 6 | 876 | 69 | |
| AF193017.1 | - | 5 - 6 | 885 | 69 | ||
| AK013330.1 | - | 5 - 6 | 927 | 69 | ||
| AK029838.1 | - | 5 - 6 | 909 | 69 | ||
| AK036866.1 | - | 5 - 6 | 911 | 69 | ||
| AK151288.1 | - | 5 - 6 | 955 | 69 | ||
| BC017527.1 | - | 5 - 6 | 876 | 4921119 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Preb
Genomic Map