Assay Details
| Gene Symbol: Dpysl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF249295.1 | - | 1 - 2 | 437 | 57 | |
| AK044866.1 | - | 1 - 2 | 145 | 57 | ||
| BC103789.1 | - | 1 - 2 | 100 | 57 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF249295.1, BC103789.1 transcripts.
Set Membership: |
|

Dpysl5
Genomic Map