Assay Details
| Gene Symbol: Ccr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009912.4 | NP_034042.3 | 1 - 2 | 396 | 149 | |
| GenBank mRNA | AK031109.1 | - | 1 - 2 | 397 | 149 | |
| AK036597.1 | - | 1 - 2 | 98 | 149 | ||
| AK036690.1 | - | 1 - 2 | 396 | 149 | ||
| AK076275.1 | - | 1 - 2 | 271 | 149 | ||
| BC011092.1 | - | 1 - 2 | 99 | 3992755 | 149 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK031109.1, AK036597.1, AK036690.1, AK076275.1, BC011092.1, NM_009912.4 transcripts.
Set Membership: |
|

Ccr1
Genomic Map