Assay Details
| Gene Symbol: Cenpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040313.1 | - | 1 - 2 | 202 | 60 | |
| AK041138.1 | - | 1 - 2 | 195 | 60 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cenpa| Gene Symbol: Cenpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040313.1 | - | 1 - 2 | 202 | 60 | |
| AK041138.1 | - | 1 - 2 | 195 | 60 | ||
Set Membership: |
|