Assay Details
| Gene Symbol: Apbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201414.1 | NP_001188343.1 | 5 - 6 | 1467 | 88 | |
| GenBank mRNA | AK032770.1 | - | 5 - 6 | 1270 | 88 | |
More Information
Set Membership: |
|

Apbb2
Genomic Map