Assay Details
| Gene Symbol: Corin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122756.1 | NP_001116228.1 | 13 - 14 | 1945 | 74 | |
| NM_016869.3 | NP_058565.2 | 13 - 14 | 2102 | 74 | ||
| GenBank mRNA | AB013874.1 | - | 13 - 14 | 2102 | 74 | |
| BC093485.1 | - | 13 - 14 | 1878 | 30648458 | 74 | |
| BC138339.1 | - | 13 - 14 | 2085 | 8861359 | 74 | |
| BC138340.1 | - | 13 - 14 | 2085 | 8861360 | 74 | |
| BC145243.1 | - | 13 - 14 | 2085 | 9053795 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Corin
Genomic Map